Order Entry
Puerto Rico
ContactUsLinkComponent
Riboprobe Combination System-SP6/T7 RNA Polymerase, 1 system, Promega
Riboprobe Combination System-SP6/T7 RNA Polymerase, 1 system, Promega
Catalog # PAP1460
CAS Number:  
undefined
Riboprobe Combination System-SP6/T7 RNA Polymerase, 1 system, Promega
Catalog # PAP1460
Supplier Number:  P1460
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Riboprobe Combination System-SP6/T7 RNA Polymerase, 1 system
  • Size:
    1 system
  • Storage Conditions:
    -30°C to -10°C
  • Cat. No.:
    PAP1460
  • Supplier no.:
    P1460

Specifications

About this item

The Riboprobe Systems are designed to prepare high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts.

  • Produce High-Specific-Activity RNA Probes or Defined RNA Transcripts
  • Choose from combination systems with T3/T7 or SP6/T7 RNA polymerases
  • SP6, T7 and T3 RNA Polymerases are extremely promoter-specific, allowing production of virtually homogeneous RNA using plasmid DNA as a template
  • Allows synthesis of RNA corresponding to the coding or noncoding strand of cloned DNA from a single plasmid construct

The Riboprobe Combination Systems are designed for in vitro preparation of high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts. The Riboprobe Combination Systems include the RNA polymerases, all of the required reagents (excluding radioisotope) for performing transcription reactions in vitro and RQ1 RNase-Free DNase (Cat.# M6101) for removal of the template following transcription.