Ribo m7G Cap Analog, A254 units, Promega
Supplier: Promega Corporation
Certificates
About this item
This methylated ribonucleotide can be incorporated onto the 5'-end of in vitro transcripts and simulates the cap structure found on most eukaryotic mRNA molecules.
- Enhances Translation Efficiency in Reticulocyte-Based Reactions
- Protects RNA from intracellular digestion
- Can be used with Riboprobe(R) or RiboMAX™ RNA production systems
- Supplied at a concentration of 40mM
The Ribo m(7)G Cap Analog is a modified ribonucleotide with the structure (m(7)G(5')ppp(5')G). This methylated ribonucleotide can be incorporated onto the 5'-end of transcripts synthesized in vitro and simulates the 7-methyl guanosine 5'-cap structure found on most eukaryotic mRNA molecules.
Caution: Store at –20°C