Red Blood Cell Lysis Solution (CLB), 200ml, Promega
Catalog # PAZ3141
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:Red Blood Cell Lysis Solution (CLB)
- Size:200 ml
- Storage Temperature:15°C to 30°C
- Cat. No.:PAZ3141
- Supplier no.:Z3141
Specifications
About this item
Component of the SV Total RNA Isolation and PureYield RNA Midiprep Systems.
- Component of the SV Total RNA Isolation and PureYield RNA Midiprep Systems.
Red Blood Cell Lysis Solution (CLB) is a component of the SV Total RNA Isolation and PureYield RNA Midiprep Systems.