Order Entry
Puerto Rico
ContactUsLinkComponent
rATP, 10 mM, 0.5 ml, Promega
rATP, 10 mM, 0.5 ml, Promega
Catalog # PAP1132
CAS Number:  
undefined
rATP, 10 mM, 0.5 ml, Promega
Catalog # PAP1132
Supplier Number:  P1132
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    rATP, 10mM, 0.5ml
  • Size:
    0.5ml
  • Storage Conditions:
    -30°C to -10°C
  • Cat. No.:
    PAP1132
  • Supplier no.:
    P1132

Specifications

About this item

High-quality rNTPs. Provided at concentrations of 10mM or 100mM in water at neutral pH.

  • High-Quality Ribonucleoside Triphosphates (rNTPs)
  • 400µl rNTP supplied at 10mM or 100mM in water at neutral pH
  • Qualified for use with the Riboprobe(R) and HeLaScribe(R) Systems
  • Purity verified by HPLC

Ribonucleotide Triphosphates (rNTPs) are provided in individual tubes and qualified for use with the Riboprobe and HeLaScribe Systems. The rNTPs are supplied in nuclease-free water. Purity is verified by HPLC analysis.