Order Entry
Puerto Rico
ContactUsLinkComponent
Random Primers, 20 µg, Promega
Random Primers, 20 µg, Promega
Catalog # PAC1181
CAS Number:  
undefined
Random Primers, 20 µg, Promega
Catalog # PAC1181
Supplier Number:  C1181
CAS Number:  

Specifications

  • Description:
    Random Primers
  • Size:
    20 μg
  • Storage Conditions:
    -30°C to -10°C
  • Cat. No.:
    PAC1181
  • Supplier no.:
    C1181

Specifications

About this item

Use for first-strand cDNA synthesis with a reverse transcriptase.

  • Streamline Universal cDNA Synthesis
  • High-quality DNA hexamers of randomized sequence

Random Primers can be used for first-strand cDNA synthesis and cloning; they also are available as components of the Universal Riboclone cDNA Synthesis System (Cat.# C4360) and Reverse Transcription System (Cat.# A3500). The primers are random hexadeoxynucleotides.