Order Entry
Puerto Rico
ContactUsLinkComponent
pUC/M13 Primer, Reverse (17mer), 2 µg, Promega
pUC/M13 Primer, Reverse (17mer), 2 µg, Promega
Catalog # PAQ5401
CAS Number:  
undefined
pUC/M13 Primer, Reverse (17mer), 2 µg, Promega
Catalog # PAQ5401
Supplier Number:  Q5401
CAS Number:  

Specifications

  • Description:
    pUC/M13 Primer, Reverse (17mer), 2microg
  • Size:
    2 μg
  • Storage Conditions:
    -30°C to -10°C
  • Cat. No.:
    PAQ5401
  • Supplier no.:
    Q5401

Specifications

About this item

Designed for sequencing inserts cloned into the M13 vectors and pUC plasmids. They also can be used for sequencing other lacZ-containing plasmids such as the pGEM-Z and pGEM-Zf Vectors.

  • Designed for Sequencing Inserts Cloned into the M13 Vectors and pUC Plasmids
  • Can sequence other lacZ-containing plasmids such as the pGEM(R)-Z and pGEM(R)-Zf Vectors
  • Supplied at a concentration of 10microg/ml

The pUC/M13 Primers are designed for sequencing inserts cloned into the M13 vectors and pUC plasmids. These primers also can be used for sequencing other lacZ-containing plasmids such as the pGEM-Z and pGEM-Zf Vectors. The primers are purified by gel electrophoresis or HPLC. Primer Sequences: [Forward (17mer)] 5'-d(GTTTTCCCAGTCACGAC)-3', [Reverse (17mer)] 5'-d(CAGGAAACAGCTATGAC)-3', [Reverse (22mer)] 5'-d(TCACACAGGAAACAGCTATGAC)-3', [Forward (24mer)] 5'-d(CGCCAGGGTTTTCCCAGTCACGAC)-3'.