Order Entry
United States
ContactUsLinkComponent
Markers Lambda Ladders, 40 to 60 lanes, Promega
Markers Lambda Ladders, 40 to 60 lanes, Promega
Catalog # PAG3011
CAS Number:  
undefined
Markers Lambda Ladders, 40 to 60 lanes, Promega
Catalog # PAG3011
Supplier Number:  G3011
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ProMega-Markers Lambda Ladders, 40-60 lanes
  • Size:
    40-60 lanes
  • Environmentally Preferable:
  • Storage Temperature:
    2°C to 10°C
  • Cat. No.:
    PAG3011
  • Supplier no.:
    G3011

Specifications

About this item

50-1,000kb; 20 bands. Concatamer of lambda phage DNA. Embedded in dye-colored, 1% agarose string molds in 50mM EDTA at 0.5ug/5mm.

  • λ Phage DNA Concatemerized into Multimers of 50-800kb+
  • Embedded in dye-colored, 0.5% agarose string molds in 50mM EDTA
  • DNA concentration is 0.5microg/5mm
  • Certified for use as markers in pulsed-field gel electrophoresis

ProMega-Markers Lambda Ladders are prepared by concatemerization of lambda phage DNA into multimers ranging in size from 50kb to 800kb and up, with each multimer, or rung, of the 20-step ladder differing in size by one lambda genome (approximately 48.5kb). The ladders are embedded in dye-colored, 0.5% agarose string molds in 50mM EDTA. The ladders are not intended for use in quantitative analysis.