Order Entry
Puerto Rico
ContactUsLinkComponent
PI3K (p110delta/p85alpha)
PI3K (p110delta/p85alpha)
Catalog # PAV1771
CAS Number:  
undefined
PI3K (p110delta/p85alpha)
Catalog # PAV1771
Supplier Number:  V1771
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    200 μl
  • Storage Conditions:
    LESS THAN −65 °C
  • Enzyme Name:
    PI3K (p110delta/p85alpha)
  • Cat. No.:
    PAV1771
  • Supplier no.:
    V1771

Specifications

About this item

The PI3K lipid kinase enzymes are used with ADP-Glo KInase Assay System and Lipid Kinase Substrates in a homogeneous, add-mix-read lipid kinase assay.

  • Purified PI3K Class I Lipid Kinases
  • Use with ADP-Glo™ Assay and PI3K Substrates in lipid kinase activity assays

Phosphatidylinositol (PI) and its phosphorylated derivates, collectively called phosphoinositides, are important second messengers that are critical as signaling molecules and for cellular membrane remodeling. The PI3K lipid kinase enzymes are used with ADP-Glo KInase Assay System and Lipid Kinase Substrates in a homogeneous, add-mix-read lipid kinase assay.