ContactUsLinkComponent
PI3K (p110alpha/p85alpha)
  PAV1721
 :  
undefined
PI3K (p110alpha/p85alpha)
  PAV1721
 :  V1721
 :  
  • Size:
    200 μl
  • Storage Conditions:
    LESS THAN −65 °C
  • Enzyme Name:
    PI3K (p110alpha/p85alpha)
  • Cat. No.:
    PAV1721
  • Supplier no.:
    V1721
The PI3K lipid kinase enzymes are used with ADP-Glo KInase Assay System and Lipid Kinase Substrates in a homogeneous, add-mix-read lipid kinase assay.
  • Purified PI3K Class I Lipid Kinases
  • Use with ADP-Glo™ Assay and PI3K Substrates in lipid kinase activity assays
Phosphatidylinositol (PI) and its phosphorylated derivates, collectively called phosphoinositides, are important second messengers that are critical as signaling molecules and for cellular membrane remodeling. The PI3K lipid kinase enzymes are used with ADP-Glo KInase Assay System and Lipid Kinase Substrates in a homogeneous, add-mix-read lipid kinase assay.