Order Entry
Puerto Rico
ContactUsLinkComponent
 
PI:3PS Lipid Kinase Substrate, 0.5mg, 0.5ml, Promega
  PAV1711
 :  
undefined
PI:3PS Lipid Kinase Substrate, 0.5mg, 0.5ml, Promega
  PAV1711
 :  V1711
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    PI:3PS Lipid Kinase Substrate, 0.5mg, 0.5ml
  • Size:
    0.5ml
  • Storage Temperature:
    LESS THAN -65°C
  • Cat. No.:
    PAV1711
  • Supplier no.:
    V1711

 

 

The lipid kinase substrates PIP2:3PS and PI:3PS are standalone substrates that are also part of the PI3K-Glo kinase profiling system.

  • Phosphoinositide-3 KInase (PI3K) Substrates
  • Use in screening assays with purified PI3K Class I lipid kinases and ADP-Glo™ Kinase Assay
  • PIP2:3PS supplied at 0.25mg in 0.25ml
  • PI:3PS supplied at 0.5mg in 0.5ml

The lipid kinase substrates PIP2:3PS and PI:3PS are standalone substrates that are part of the PI3K-Glo kinase profiling system. Promega lipid kinase enzymes, substrates and detection systems provide a complete set of reagents for performing phosphoinositide lipid kinase (PIK) reactions using a luminescent ADP-detection platform, the ADP-Glo Kinase Assay. The lipid kinase reaction is performed by incubating lipid substrate (PI:3PS or PIP2:3PS) with a recombinant enzyme and ATP, and the kinase activity is measured using the ADP-Glo Kinase Assay.