To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:PhiX174, RF DNA, 50microg
- Size:50 μg
- Environmentally Preferable:
- Storage Temperature:-30°C to -10°C
- Cat. No.:PAD1531
- Supplier no.:D1531
Specifications
About this item
phiX174 replicative form (RF) is a double-stranded circular DNA molecule of 5,386 bases. Used in assays of restriction enzymes for the presence of nickase activity.
- Double-Stranded Circular DNA Molecule Used to Detect Restriction Enzyme Assay Nickase Activity
- Supplied at a concentration of 1mg/ml
The icosahedral bacteriophage PhiX174 replicative form (RF) is a double-stranded circular DNA molecule of 5,386 bases. Restriction enzyme-digested PhiX174 DNA generates molecular weight size markers used in gel analysis of nucleic acids. PhiX174 DNA is often used in the assays of restriction enzymes for the presence of nickase activity.