Order Entry
Puerto Rico
ContactUsLinkComponent
 
DNA (Deoxyribonucleic acid) pGEM Beta-Gal , control DNA
Catalog # PAL4731
Supplier:  Avantor
CAS Number:  
undefined
DNA (Deoxyribonucleic acid) pGEM Beta-Gal , control DNA
Catalog # PAL4731
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAL4731
  • Supplier no.:
    L4731

Specifications

About this item

 :
Storage Temperature:Freezer
CAS Number:9007-49-2
 :73049-39-5
MDL Number:MFCD00130922