To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:BenchTop pGEM DNA Markers, 250microl
- Size:250 μl
- Environmentally Preferable:
- Storage Temperature:-30°C to 30°C
- Cat. No.:PAG7521
- Supplier no.:G7521
Specifications
About this item
36-2,645bp; 15 bands. Can be stored at room temperature. Contains dye marker, ready to load. Combination of three separate digests of pGEM(R)-3 Vector.
- Fifteen DNA Fragments of 36-2,645bp, Combined From Three Separate Digests of pGEM(R)-3 Vector
- Store at room temperature (22-25°C)
- Premixed with loading buffer; ready to load onto agarose gels
- Load 5microl/lane
The BenchTop DNA Markers offer the convenience of storage at room temperature (22-25°C) as well as the capability of direct loading onto agarose gels. The BenchTop DNA Markers are supplied in a stabilizing solution of 1X Blue/Orange Loading Dye, which circumvents any requirements for further manipulation. Fifteen phenol-extracted, ethanol-precipitated DNA fragments ranging in size from 36bp to 2,645bp. These unique markers are generated from separate digests of pGEM(R)-3 Vector DNA with HinfI, RsaI and SinI later combined to form the markers. Load 5microl/lane.