- Description:P450-Glo CYP2C9 Screening System, 1,000 assays
- Size:1,000 assays
- Storage Temperature:LESS THAN -65°C
- Cat. No.:PAV9790
- Supplier no.:V9790
The P450-Glo Screening Systems provide a complete set of reagents for performing luminescent cytochrome P450 assays
- A Luminescent Method and Screening System for Measuring CYP2C9 Activity
- Screening system includes complete set of reagents
- Luminescent format eliminates need for time-consuming analyses
- Broad dynamic range and low background
The P450-Glo Screening Systems provide a complete set of reagents for performing luminescent cytochrome P450 assays. The systems include a membrane preparation containing recombinant human cytochrome P450 enzyme, a luminogenic cytochrome P450 substrate appropriate for the enzyme, an NADPH Regeneration System, reaction buffer, Luciferin Detection Reagent and Luciferin-Free Water. The membranes are prepared from baculovirus-infected insect cells and contain human cytochrome P450 and P450 reductase (and cytochrome b5 for CYP2C9 and CYP3A4). The P450-Glo Screening Systems also contain a membrane fraction devoid of cytochrome P450 activity as a negative control. The assays are ideal for testing the effects of drugs and new chemical entities on cytochrome P450 enzyme activities. The cytochrome P450 reaction is performed by incubating a luminogenic cytochrome P450 substrate with a cytochrome P450 enzyme and the NADPH Regeneration System. The luminogenic P450-Glo Substrates are derivatives of beetle luciferin ((4S)-4,5-dihydro-2-(6-hydroxybenzothiazolyl)-4-thiazolecarboxylic acid or D-luciferin), a substrate of firefly luciferase. The P450-Glo Substrates do not react with luciferase but are converted by cytochrome P450 to luciferin, which in turn reacts with luciferase to produce light. Light is used to monitor cytochrome P450 activity because the amount of light produced is directly proportional to the amount of D-luciferin produced by cytochrome P450. The new P450-Glo CYP3A4 Screening System with Luciferin-IPA contains our latest 3A4 substrate and is the most sensitive P450-Glo 3A4 substrate available. Luciferin-IPA displays the widest range of inhibition profiles of all the P450-Glo CYP3A4 substrates. Dimethyl sulfoxide (DMSO), a common solvent used to solubilize chemical compounds, can significantly inhibit the activity of the 3A4 isoform of cytochrome P450, even at low concentrations (<0.1%). The P450-Glo CYP3A4 Screening System (Luciferin-PPXE) DMSO-Tolerant Assay is specifically designed to tolerate DMSO in the 3A4 reaction. The assay exhibits little to no change in the signal-to-background ratio in the presence of 0.2% DMSO compared to a no-DMSO control. After the cytochrome P450 reaction has been performed, the reconstituted Luciferin Detection Reagent is added. This reagent simultaneously stops the cytochrome P450 reaction and initiates a stable glow-type luminescent signal. The glow-type reaction produces a stable signal and eliminates the need for strictly timed luminescence detection. Protocols are configured for multiwell plate formats but can be easily adapted for single-tube applications.