Order Entry
Puerto Rico
ContactUsLinkComponent
P450-Glo™ CYP1A2 Screening System, Promega
P450-Glo™ CYP1A2 Screening System, Promega
Catalog # PAV9770
CAS Number:  
undefined
P450-Glo™ CYP1A2 Screening System, Promega
Catalog # PAV9770
Supplier Number:  V9770
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    P450-Glo CYP1A2 Screening System, 1,000 assays
  • Size:
    1,000 assays
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV9770
  • Supplier no.:
    V9770

Specifications

About this item

The P450-Glo CYP1A2 Assay and Screening Systems provide a complete set of reagents for performing luminescent cytochrome P450 assays

  • A Luminescent Assay and Screening System for Measuring CYP1A2 Activity
  • Screening system includes a complete set of reagents
  • Luminescent format eliminates need for time-consuming analyses
  • Broad dynamic range and low background

The P450-Glo Screening Systems provide a complete set of reagents for performing luminescent cytochrome P450 assays. The systems include a membrane preparation containing recombinant human cytochrome P450 enzyme, a luminogenic cytochrome P450 substrate appropriate for the enzyme, an NADPH Regeneration System, reaction buffer, Luciferin Detection Reagent and Luciferin-Free Water. The membranes are prepared from baculovirus-infected insect cells and contain human cytochrome P450 and P450 reductase (and cytochrome b5 for CYP2C9 and CYP3A4). The P450-Glo Screening Systems also contain a membrane fraction devoid of cytochrome P450 activity as a negative control. The assays are ideal for testing the effects of drugs and new chemical entities on cytochrome P450 enzyme activities. The cytochrome P450 reaction is performed by incubating a luminogenic cytochrome P450 substrate with a cytochrome P450 enzyme and the NADPH Regeneration System. The luminogenic P450-Glo Substrates are derivatives of beetle luciferin ((4S)-4,5-dihydro-2-(6-hydroxybenzothiazolyl)-4-thiazolecarboxylic acid or D-luciferin), a substrate of firefly luciferase. The P450-Glo Substrates do not react with luciferase but are converted by cytochrome P450 to luciferin, which in turn reacts with luciferase to produce light. Light is used to monitor cytochrome P450 activity because the amount of light produced is directly proportional to the amount of D-luciferin produced by cytochrome P450. The new P450-Glo CYP3A4 Screening System with Luciferin-IPA contains our latest 3A4 substrate and is the most sensitive P450-Glo 3A4 substrate available. Luciferin-IPA displays the widest range of inhibition profiles of all the P450-Glo CYP3A4 substrates. Dimethyl sulfoxide (DMSO), a common solvent used to solubilize chemical compounds, can significantly inhibit the activity of the 3A4 isoform of cytochrome P450, even at low concentrations (<0.1%). The P450-Glo CYP3A4 Screening System (Luciferin-PPXE) DMSO-Tolerant Assay is specifically designed to tolerate DMSO in the 3A4 reaction. The assay exhibits little to no change in the signal-to-background ratio in the presence of 0.2% DMSO compared to a no-DMSO control. After the cytochrome P450 reaction has been performed, the reconstituted Luciferin Detection Reagent is added. This reagent simultaneously stops the cytochrome P450 reaction and initiates a stable glow-type luminescent signal. The glow-type reaction produces a stable signal and eliminates the need for strictly timed luminescence detection. Protocols are configured for multiwell plate formats but can be easily adapted for single-tube applications.