ContactUsLinkComponent
One-Way Luer-Lok Stopcocks for Nucleic Acid Purification Systems, Promega®
One-Way Luer-Lok Stopcocks for Nucleic Acid Purification Systems, Promega®
Catalog # PAA7261
undefined
One-Way Luer-Lok Stopcocks for Nucleic Acid Purification Systems, Promega®
Catalog # PAA7261
Supplier Number:  A7261
Specifications
  • Description:
    One-Way Luer-Lok Stopcocks
  • Size:
    10 each
  • Cat. No.:
    PAA7261
  • Supplier no.:
    A7261
Specifications
About this item
One-Way Luer-Lok stopcocks vacuum manifold for nucleic acid purification system. Spinlock adapters on the columns from the Wizard DNA purifications systems can be attached to the upper row of stopcocks for easy manipulation of the stopcock valves.