ContactUsLinkComponent
Oligo(dT)15 Primer, 20 µg, Promega
Oligo(dT)15 Primer, 20 µg, Promega
  PAC1101
 :  
undefined
Oligo(dT)15 Primer, 20 µg, Promega
  PAC1101
 :  C1101
 :  
  • Description:
    Oligo(dT)15 Primer
  • Size:
    20 μg
  • Storage Conditions:
    -30°C to -10°C
  • Cat. No.:
    PAC1101
  • Supplier no.:
    C1101
Use for first-strand cDNA synthesis with a reverse transcriptase.
  • Simplify Your First-Strand cDNA Synthesis
  • Primer for reverse transcription of polyadenylated RNAs, such as mRNAs
  • Provided at 500µg/ml
Oligo(dT)15 Primer is suitable for use as a primer for first-strand cDNA synthesis with a reverse transcriptase. The primer hybridizes to the poly(A) tail of mRNA.