DNA (Deoxyribonucleic acid) (Mouse genomic)
Catalog # PAG3091
Supplier: Promega Corporation
CAS Number:
Specifications
- Size:100 μg
- Cat. No.:PAG3091
- Supplier no.:G3091
Specifications
About this item
:
CAS Number:9007-49-2
:73049-39-5
MDL Number:MFCD00130922