ContactUsLinkComponent
Maxwell RSC IQ/OQ Combination Package, 1 each, Promega
Maxwell RSC IQ/OQ Combination Package, 1 each, Promega
Catalog # PASA1349
undefined
Maxwell RSC IQ/OQ Combination Package, 1 each, Promega
Catalog # PASA1349
Supplier Number:  SA1349
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Maxwell RSC IQ/OQ Combination Package, 1 each
  • Environmentally Preferable:
  • Cat. No.:
    PASA1349
  • Supplier no.:
    SA1349
Specifications
About this item
Maxwell RSC Installation Qualification and Operational Qualification Products.
  • Choose the Installation and Operational Qualification Products that Best Suit Your Needs
  • The Installation Qualification (IQ) includes a series of instrument checks, delivers written documentation of functionality and demonstrates that everything ordered with the instrument is supplied and installed
  • The Operational Qualification (OQ) service product demonstrates that the instrument functions according to its operational specifications
  • The IQ/OQ Package combines both services
Maxwell RSC Installation Qualification and Operational Qualification Products, Cat.# SA1347, SA1348, SA1349: The Installation Qualification service product includes a series of formal instrument checks, delivers written documentation of instrument functionality and demonstrates that everything ordered with the instrument is supplied and installed at the customer’s laboratory. This service product must be delivered by a Promega representative who is certified to perform the Installation Qualification. The service product involves a site visit to perform: Installation by qualified Promega personnel; Inspection of shipping containers, instrument and accessories; Comparison of items received vs. items on purchase order; Inspection of laboratory conditions (power, etc.); Review all hazards and precautions with users; Confirmation/installation of correct firmware version; Testing of instrument run; Recording and documenting installation and actions. The Operational Qualification (OQ) service product demonstrates that the instrument functions according to its operational specifications. This service product must be delivered by a Promega representative who is certified to perform the Operational Qualification. The service product involves a site visit to perform: Running operational verification tests; Documenting all calibration and test results; Training user to operate the instrument; Trainings users to use the log book; Completing customer-specific log book, instrument sticker and OQ documentation.