About this item
The MagaZorb DNA Kit provides an easy, fast and cost-effective technique for isolating PCR-quality DNA.
- Fast Isolation of PCR-quality DNA from Various Samples
- All reagents provided, no reagent preparation required
- Magnetic purification method increases throughput and reproducibility
- No organic solvents
The MagaZorb DNA Kit provides an easy, fast and cost-effective technique for isolating PCR-quality DNA. Using one simple protocol, a high yield of purified DNA can be isolated from a wide variety of sources including whole blood (fresh or frozen, citrate-, heparin- or EDTA-treated), buffy coat, leukocytes, milk, seminal fluid, dried blood spots, cultured cells, tissue (fresh, frozen or formalin-fixed paraffin-embedded), saliva, urine, stool, hair, buccal swabs and vaginal swabs. The 20-Position Microcentrifuge Tube Magnetic Separator (Cat.# CD4002) utilizes a microcentrifuge tube rack that can be removed from the high-strength magnets for wash steps or incubation in a water bath. The rack is designed to hold the microcentrifuge tubes so that they will not fall out even when turned upside down, and it can withstand temperatures of up to 80 degrees C for convenient manipulation of sample tubes. Please note that the magnets in the 20-Position Microcentrifuge Tube Magnetic Separator are designed specifically for use with the MagaZorb DNA Kit; separation may not work with other particles.
Specifications
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Blood (dried) on filter cardsBloodCultured cellsFFPE tissuesHuman/animal cells and tissues
- Target molecule:DNA
- Storage Conditions:15°C to 30°C
- Description:MagaZorb DNA Mini-Prep Kit