Specifications
- Description:Lambda DNA/EcoRI Markers, 100microg
- Size:100 μg
- Storage Temperature:-30°C to -10°C
- Cat. No.:PAG1721
- Supplier no.:G1721
Specifications
About this item
Generated by digesting lambda DNA to completion with EcoRI, resulting in six DNA fragments of 3,530-21,226bp.
- Six DNA Fragments of 3,530-21,226bp
- Load 1microl/lane
- Supplied at a concentration of 0.5µg/microl
- Supplied with 6X Blue/Orange Loading Dye
The Conventional DNA Digest Markers are created by digesting either lambda DNA, PhiX174 replicative form DNA, or plasmids to completion with one or more restriction enzymes. The enzymes are heat-inactivated, and the DNA fragments are either phenol-extracted, then ethanol-precipitated or just ethanol-precipitated. The precipitated fragments are resuspended in storage buffer. The markers are not intended for quantitative analysis. Each marker is supplied with a tube of 6X Blue/Orange Loading Dye. Lambda DNA/EcoRI Markers: Six ethanol-precipitated DNA fragments ranging in size from 3,530bp to 21,226bp. Recommended loading: 1microl/lane.