Order Entry
Puerto Rico
ContactUsLinkComponent
Lambda DNA/EcoRI + HindIII Markers, 100 µg, Promega
Lambda DNA/EcoRI + HindIII Markers, 100 µg, Promega
Catalog # PAG1731
CAS Number:  
undefined
Lambda DNA/EcoRI + HindIII Markers, 100 µg, Promega
Catalog # PAG1731
Supplier Number:  G1731
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Lambda DNA/EcoRI + HindIII Markers, 100microg
  • Size:
    100 μg
  • Storage Temperature:
    -30°C to -10°C
  • Cat. No.:
    PAG1731
  • Supplier no.:
    G1731

Specifications

About this item

Created by digesting lambda DNA to completion with EcoRI and HindIII to generate 13 fragments of 125-21,226bp.

  • Thirteen DNA Fragments of 125-21,226bp
  • Load 1microl/lane
  • Supplied at a concentration of 0.5µg/microl
  • Supplied with 6X Blue/Orange Loading Dye

The Conventional DNA Digest Markers are created by digesting either lambda DNA, PhiX174 replicative form DNA, or plasmids to completion with one or more restriction enzymes. The enzymes are heat-inactivated, and the DNA fragments are either phenol-extracted, then ethanol-precipitated or just ethanol-precipitated. The precipitated fragments are resuspended in storage buffer. The markers are not intended for quantitative analysis. Each marker is supplied with a tube of 6X Blue/Orange Loading Dye. Lambda DNA/EcoRI + HindIII Markers: Thirteen ethanol-precipitated DNA fragments ranging in size from 125bp to 21,226bp. Recommended loading: 1microl/lane