Order Entry
Puerto Rico
ContactUsLinkComponent
Lambda DNA, 250 µg, Promega
Lambda DNA, 250 µg, Promega
Catalog # PAD1501
CAS Number:  
undefined
Lambda DNA, 250 µg, Promega
Catalog # PAD1501
Supplier Number:  D1501
CAS Number:  

Specifications

  • Description:
    Lambda DNA, 250microg
  • Size:
    250 μg
  • Storage Temperature:
    -30°C to -10°C
  • Cat. No.:
    PAD1501
  • Supplier no.:
    D1501

Specifications

About this item

Generates molecular weight size markers used in gel analysis of nucleic acids; nucleotide sequence has been determined.

  • Generates molecular weight size markers used in gel analysis of nucleic acids; nucleotide sequence has been determined.

Lambda DNA cl857 Sam7 is isolated from infected E. coli strain W3350. Restriction enzyme-digested lambda DNA (48,502bp) may be used as a molecular weight size marker in gel analysis of nucleic acids. Lambda DNA is also a commonly used substrate in restriction enzyme activity assays. The nucleotide sequence has been determined.