Order Entry
Puerto Rico
ContactUsLinkComponent
JM109 Competent Cells, >10(8) cfu/μg, 1 ml, Promega
JM109 Competent Cells, >10(8) cfu/μg, 1 ml, Promega
  PAL2001
 :  
undefined
JM109 Competent Cells, >10(8) cfu/μg, 1 ml, Promega
  PAL2001
 :  L2001
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    JM109 Competent Cells, >10(8)cfu/ug, 1ml
  • Size:
    1ml
  • Storage Conditions:
    Store at –70°C
  • Cat. No.:
    PAL2001
  • Supplier no.:
    L2001

 

 

JM109 is a K strain that is recA- and endA- to minimize recombination and improve the quality of plasmid DNA. In addition, the cells carry the F´ episome, which allows blue/white screening.

  • Ready-to-Use Competent E. coli Cells for Recombinant Cloning and Subcloning
  • Cells are ready to use—no preparation time necessary
  • Transformation efficiencies guaranteed
  • No worries about undesirable recombination events (recA- mutation), and no nuclease carryover in miniprep DNA (endA- mutation)

JM109 is a K strain that is recA- and endA- to minimize recombination and improve the quality of plasmid DNA. In addition, the cells carry the F´ episome, which allows blue/white screening. JM109 Competent Cells are available for convenient transformation in two efficiencies: High Efficiency at greater than 10(8)cfu/ug and Subcloning Efficiency at greater than 10(7)cfu/ug. JM109 Competent Cells are prepared according to a modified procedure of Hanahan. JM109 Genotype: endA1, recA1, gyrA96, thi, hsdR17 (r(k)-, m(k)+), relA1, supE44, Delta(lac-proAB), [F' traD36, proAB, laqI(q)ZDeltaM15].