About this item
Recombinant full-length human IRAK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Interleukin-1 receptor-associated kinase 4 is an important mediator in the signal transduction of Toll-like receptor and IL1R family members.
- Easily Screen and Profile IRAK4 Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human IRAK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Interleukin-1 receptor-associated kinase 4 (IRAK4) is an important mediator in the signal transduction of Toll-like receptor (TLR) and IL1R family members. IRAK4 is involved in the Toll-like receptor signaling pathway leading to Apoptosis. IRAK4 has molecular functions like protein binding, ATP binding, kinase activity and magnesium ion binding. Toll/IL-1 receptor family members like IRAK4 are central components of host defense mechanisms in a variety of species. One well conserved element in their signal transduction is the Ser/Thr kinase activity, which couple early signaling events in a receptor complex at the plasma membrane to larger signalosomes in the cytosol. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: IRAK4, 10ug (Human, recombinant full-length). MW: ~81kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. IRAK4 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/51135/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Temperature:LESS THAN -65°C