About this item
Recombinant human InsR (amino acids 1011-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. InsR is the insulin receptor tyrosine kinase that is involved in insulin signaling.
- Easily Screen and Profile InsR Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human InsR (amino acids 1011-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. InsR is the insulin receptor tyrosine kinase that is involved in insulin signaling. InsR is post-translationally cleaved into two chains, alpha and beta, that are covalently linked. Binding of insulin to the InsR stimulates glucose uptake. Insulin receptor signaling helps to maintain fuel homeostasis and prevent diabetes. Studies have shown that a conditional knockout of insulin receptor substrate 2 (IRS2) in mouse pancreas beta cells and parts of the brain---including the hypothalamus---increased appetite, lean and fat body mass, linear growth, and insulin resistance that progressed to diabetes. InsR signaling also increases the regeneration of adult beta cells and the central control of nutrient homeostasis. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: InsR, 10ug (Human, recombinant; amino acids 1011-end). MW: ~70kDa. Substrate: Axltide (KKSRGDYMTMQIG); derived from the mouse Insulin receptor substrate 1 (amino acid 979-989). Other: Reaction Buffer, DTT, MnCl2. InsR NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/3643/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Specifications
- Storage Temperature:LESS THAN -65°C