Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DNA (Deoxyribonucleic acid)
DNA (Deoxyribonucleic acid) (from herring sperm)
DNA (Deoxyribonucleic acid) (from herring sperm)
DNA (Deoxyribonucleic acid) (from herring sperm)
Supplier:
Promega Corporation
Certificates
Expand image
DNA (Deoxyribonucleic acid) (from herring sperm)
Supplier:
Promega Corporation
Certificates
About this item
CAS Number:
9007-49-2
:
73049-39-5
MDL Number:
MFCD00130922
Storage Temperature:
Freezer
Related Information
Documentation
Certificates
Search by Lot Number
Documentation