Order Entry
Puerto Rico
ContactUsLinkComponent
HB101 Competent Cells, >10(8)cfu/ug, 1ml, Promega
HB101 Competent Cells, >10(8)cfu/ug, 1ml, Promega
Catalog # PAL2011
CAS Number:  
undefined
HB101 Competent Cells, >10(8)cfu/ug, 1ml, Promega
Catalog # PAL2011
Supplier Number:  L2011
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    HB101 Competent Cells, >10(8)cfu/ug, 1ml
  • Size:
    1ml
  • Storage Conditions:
    Store at –70°C
  • Cat. No.:
    PAL2011
  • Supplier no.:
    L2011

Specifications

About this item

HB101 cells are recA- to minimize recombination events and contain a restriction-minus background (hsdS20) to allow better representation when cloning methylated DNA. Suitable for vectors that do not require alpha-complementation for blue/white screening.

  • Ready-to-Use Competent E. coli Cells for Recombinant Cloning and Subcloning
  • Cells are ready to use—no preparation time necessary
  • Transformation efficiencies guaranteed
  • No worries about undesirable recombination events (recA- mutation)

HB101 cells are recA-, which minimizes recombination events, and contain a restriction-minus background (hsdS20) that allows better representation when cloning methylated DNA. HB101 Competent Cells are suitable for vectors that do not require alpha-complementation for blue/white screening. For researchers doing more than one transformation, competent cells are available in standard format (200ul aliquots). For added convenience, single-use competent cells (50ul aliquots) also are offered. HB101 Competent Cells are prepared according to a modified procedure of Hanahan. HB101 Genotype: F-, thi-1, hsdS20 (r(B)-, m(B)-), supE44, recA13, ara-14, leuB6, proA2, lacY1, galK2, rpsL20 (str(r)), xyl-5, mtl-1.