Order Entry
Puerto Rico
ContactUsLinkComponent
GoTaq PCR Core System I, 200 Reactions, Promega
GoTaq PCR Core System I, 200 Reactions, Promega
Catalog # PAM7660
CAS Number:  
undefined
GoTaq PCR Core System I, 200 Reactions, Promega
Catalog # PAM7660
Supplier Number:  M7660
CAS Number:  

Specifications

  • Description:
    GoTaq PCR Core System I, 200 reactions
  • Size:
    200 reactions
  • Storage Conditions:
    -30°C to -10°C
  • Cat. No.:
    PAM7660
  • Supplier no.:
    M7660

Specifications

About this item

Includes GoTaq DNA polymerase and PCR Nucleotide Mix, along with high-performance buffers and magnesium chloride. Core System II also includes a Positive Control Plasmid DNA and Positive Control Primers. Performance-tested in PCR.

  • Convenient Bundles Containing Taq Polymerase and Reagents for PCR
  • PCR Core System I includes GoTaq(R), Mg-free Buffers, MgCl2, and PCR Nucleotide Mix
  • PCR Core System II also includes control primers, template
  • Direct-to-gel convenience with Green GoTaq(R) Flexi Buffer

The GoTaq PCR Core Systems I and II provide all the PCR-tested components you need for specific and robust PCR amplification. You just provide the DNA template and target-specific primers. Both systems include GoTaq DNA polymerase and PCR Nucleotide Mix, along with high-performance buffers and magnesium chloride. The GoTaq PCR Core System II also includes a Positive Control Plasmid DNA and Positive Control Primers, providing increased confidence in PCR control reactions. Each system's components are performance-tested in PCR and are sufficient for 200 reactions. The GoTaq PCR Core Systems Technical Bulletin provides simple protocols and a complete troubleshooting guide to help you optimize your reactions.