GoTaq® G2 Green Master Mix, 1000 reactions, Promega
GoTaq(R) G2 Master Mixes are premixed ready-to-use solutions containing GoTaq(R) G2 DNA Polymerase, dNTPs, MgCl2 and reaction buffers at optimal concentrations for efficient amplification of a wide range of DNA templates by PCR.
- All-in-One PCR Master Mix: Just Add Template and Primers
- High-Performance GoTaq(R) G2 DNA Polymerase in a Ready-to-Use Master Mix
- Simplifies reaction setup and saves time
- Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
GoTaq G2 Green and Colorless Master Mixes are ready-to-use, green and colorless, 2X master mixes containing all necessary components for robust, reliable PCR, including the second-generation GoTaq G2 DNA Polymerase. Add template, primers and go. The second generation of GoTaq products, GoTaq G2 DNA Polymerase reliably amplifies a wide range of PCR templates and provides high-performing results due to improved manufacturing processes, increased reliability and consistency.
- Storage Conditions:-30°C to -10°C