Order Entry
Puerto Rico
ContactUsLinkComponent
GloResponse CRE-luc2P HEK293 Cell Line, 2 vials, Promega
GloResponse CRE-luc2P HEK293 Cell Line, 2 vials, Promega
Catalog # PAE8500
CAS Number:  
undefined
GloResponse CRE-luc2P HEK293 Cell Line, 2 vials, Promega
Catalog # PAE8500
Supplier Number:  E8500
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    GloResponse CRE-luc2P HEK293 Cell Line, 2 vials
  • Size:
    2 vials
  • Storage Conditions:
    BELOW -140°C
  • Cat. No.:
    PAE8500
  • Supplier no.:
    E8500

Specifications

About this item

Luciferase reporter technology is integrated into the GloResponse CRE-luc2P HEK293 Cell Line, allowing rapid analysis of cellular responses that result in modulation of CREB activities.

  • Luciferase Reporter Cell Line with CRE Response Element
  • Rapid, bioluminescent detection of any cellular response resulting in modulation of CREB activities

The GloResponse Luciferase Reporter Cell Lines contain optimized, state-of-the-art luciferase reporter technology integrated into a cell line. This allows the rapid development of a reporter assay based on the pathway of interest regulating the luciferase gene. Assays configured using the GloResponse Cell Lines are amenable for high-throughput screening. These assays typically have greater response dynamics (fold of induction) than other assay formats and good quality as indicated by the high Z' values. GloResponse Cell Lines were developed to study a variety of signaling pathways. Activators of these pathways may be native to the HEK293 cell line. Activity of non-native activators can be studied after they have been introduced by transfection. GPCRs regulate a wide-range of biological functions and are one of the most important target classes for drug discovery. GPCR signaling pathways can be categorized into three classes based on the G protein alpha-subunit involved: Gs, Gi/o and Gq. The GloResponse CRE-luc2P HEK293 Cell Line can be used to study and configure screening assays for Gs- and Gi/o-coupled GPCRs, which signal through cAMP and the cAMP Response Element (CRE). The GloResponse Cell Lines were generated by clonal selection of HEK293 cells stably transfected with pGL4-based vectors carrying specific response elements for the pathway of interest. These cell lines incorporate the improvements developed for the pGL4 family of reporter vectors for enhanced performance. The destabilized luc2P luciferase reporter is used for improved responsiveness to transcriptional dynamics. The luc2P gene is codon optimized for enhanced expression in mammalian cells, and the pGL4 plasmid backbone was engineered to reduce background reporter expression. The result is a cell line with very high induction levels when the pathway of interest is activated.