Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
dTTP, 25 µmol, Promega

dTTP, 25 µmol, Promega

dTTP, 25micromol, 25 μmol
Catalog # PAU1235
Supplier:   Promega Corporation
Supplier #:  U1235
dTTP, 25 µmol, Promega
Catalog # PAU1235
CAS Number:  
dTTP, 25 µmol, Promega

dTTP, 25micromol, 25 μmol

CAS Number:  
Price
$94.43
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

High-quality dNTPs. Provided at a concentration of 100mM in water at pH 7.5.

  • High-Quality Nucleotides for Your Workflow
  • Greater than 99% triphosphate content
  • 100mM in water at pH 7.5

High-quality deoxynucleotide triphosphates (dNTPs) are critical for the success of many key procedures such as cDNA synthesis, sequencing and labeling. Promega dNTPs have greater than 99% triphosphate content and are provided at a concentration of 100mM in water at pH 7.5.

Documents

Specifications

Specifications

  • Description:
    dTTP, 25micromol
  • Size:
    25 μmol
  • Storage Conditions:
    -30°C to -10°C
  • Cat. No.:
    PAU1235
  • Concentration:
    100 mM

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.