Order Entry
Puerto Rico
ContactUsLinkComponent
CytoTox 96 Non-Radioactive Cytotoxicity Assay, 1,000 assays, Promega
CytoTox 96 Non-Radioactive Cytotoxicity Assay, 1,000 assays, Promega
Catalog # PAG1780
CAS Number:  
undefined
CytoTox 96 Non-Radioactive Cytotoxicity Assay, 1,000 assays, Promega
Catalog # PAG1780
Supplier Number:  G1780
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    CytoTox 96 Non-Radioactive Cytotoxicity Assay, 1,000 assays
  • Size:
    1,000 assays
  • Storage Conditions:
    Store Substrate Mix and Assay Buffer at –20°C. Store LDH Positive Control, Lysis Solution (10X), and Stop Solution at 4°C.
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAG1780
  • No. of assays/kit:
    1000
  • Supplier no.:
    G1780

Specifications

About this item

Colorimetric assay for the determination of cell membrane integrity based on detection of LDH release, an indicator of cytotoxicity. Sample transfer steps are required.

  • A Colorimetric Alternative to Radioactive Cytotoxicity Assays
  • Measures release of LDH, an indicator of cytotoxicity
  • Non-radioactive, time-saving assay

The CytoTox 96 Non-Radioactive Cytotoxicity Assay is a colorimetric alternative to radioactive cytotoxicity assays. The CytoTox 96 Assay quantitatively measures lactate dehydrogenase (LDH), a stable cytosolic enzyme that is released upon cell lysis, in much the same way as [(51)Cr] is released in radioactive assays. Released LDH in culture supernatants is measured with a 30-minute coupled enzymatic assay that results in the conversion of a tetrazolium salt (INT) into a red formazan product. The amount of color formed is proportional to the number of lysed cells. Visible wavelength absorbance data are collected using a standard 96-well plate reader. The assay can be used to measure membrane integrity for cell-mediated cytotoxicity assays in which a target cell is lysed by an effector cell, or to measure lysis of target cells by bacteria, viruses, proteins, chemicals, etc.