About this item
Recombinant human c-MER (amino acids 578-872) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. c-MER is a tyrosine kinase and a member of the MER/AXL/TYRO3 receptor kinase family.
- Easily Screen and Profile c-MER Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human c-MER (amino acids 578-872) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. c-MER is a tyrosine kinase and a member of the MER/AXL/TYRO3 receptor kinase family. The c-MER gene encodes a transmembrane protein with two fibronectin type-III domains, two Ig-like C2-type domains and one tyrosine kinase domain. Functional knockout of c-MER in mice shows the mice to have macrophages deficient in the clearance of apoptotic thymocytes thereby demonstrating the critical role for c-MER in the engulfment and efficient clearance of apoptotic cells. Defects in c-MER have been associated with retinitis pigmentosa, a phenotype that can be reversed by replacement of the c-MER gene product. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: c-MER, 10ug (Human, recombinant; amino acids 578-872). MW: ~58kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT, MnCl2. c-MER NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/10461/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Temperature:LESS THAN -65°C