To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:CheckMate Mammalian Two-Hybrid System, 1 system
- Size:1 system
- Storage Temperature:-30°C to -10°C
- Cat. No.:PAE2440
- Supplier no.:E2440
Powerful system for detecting protein:protein interactions in vivo.
- Powerful System for Detecting Protein:Protein Interactions In Vivo
- Mammalian system for studying interactions in cell line of choice
- Study post-translational modifications including glycosylation, phosphorylation and acylation
Two-hybrid systems are extremely powerful methods for detecting protein:protein interactions in vivo. The basis of two-hybrid systems is the modular domains found in some transcription factors: a DNA-binding domain, which binds to a specific DNA sequence, and a transcriptional activation domain, which interacts with the basal transcriptional machinery. A transcriptional activation domain in association with a DNA-binding domain will promote the assembly of RNA polymerase II complexes at the TATA box and increase transcription. In the CheckMate Mammalian Two-Hybrid System the DNA-binding domain and the transcriptional activation domain, produced by separate plasmids, are closely associated when one protein (X) fused to a DNA-binding domain interacts with a second protein (Y) fused to a transcriptional activation domain. In this system, interaction between proteins X and Y results in transcription of a reporter gene.