ContactUsLinkComponent
Cell Lysis Solution (Genomic Purification), 1 liter, Promega
  PAA7933
 :  
undefined
Cell Lysis Solution (Genomic Purification), 1 liter, Promega
  PAA7933
 :  A7933
 :  
  • Description:
    Cell Lysis Solution (Genomic Purification), 1 liter
  • Size:
    1 liter
  • Storage Temperature:
    15 - 30 °C
  • Cat. No.:
    PAA7933
  • Supplier no.:
    A7933
An easy, solution-based method for isolation of DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria.
  • Manual, Solution-Based DNA Extraction Method
  • DNA from blood, animal and plant cells, bacteria or yeast
  • 60-minute, scalable protocol
  • Kit size based on number of isolations from whole blood, other sample types may vary
The Wizard Genomic DNA Purification Kit provides a simple, solution-based method for isolation of DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria. DNA purified with this system is suitable for a variety of applications, including amplification, digestion with restriction endonucleases and membrane hybridizations (e.g., Southern and dot/slot blots).