BSA
Catalog # PAW3841
Supplier: Promega Corporation
CAS Number:
Specifications
- Size:10 g
- Storage Conditions:15 °C to 30 °C
- Protein/Peptide Name:BSA
- Cat. No.:PAW3841
- Supplier no.:W3841
Specifications
About this item
BSA (bovine serum albumin) has been tested and qualified for optimum performance in immunoblotting applications with alkaline phosphatase antibody conjugates.
- Blot-Qualified BSA and Tween(R) 20 for Use as Blocking Agents for Antibody Detection
- Blot-Qualified BSA saturates excess sites of antibody binding on membranes for AP antibody conjugates
- Tween(R) 20 (100%) is used in immunoscreening to dissociate nonspecific interactions with an antibody probe
This BSA (bovine serum albumin) has been tested and qualified for optimum performance in immunoblotting applications with alkaline phosphatase antibody conjugates. It is shown to be alkaline phosphatase-free. Tween 20 is a nonionic detergent used as a buffer component for immunoscreening in the ProtoBlot Systems. In addition to blocking agents such as BSA, which saturate excess sites of antibody binding on membranes, this detergent acts in solution to dissociate nonspecific interactions with an antibody probe.