Reporter Lysis Buffer 5X, Promega
PAE3971
:
- Description:Reporter Lysis 5X Buffer (RLB 5X)
- Size:30 mL
- Storage Temperature:15 °C to 30 °C
- Cat. No.:PAE3971
- Supplier no.:E3971
Mild lysis reagent used with the Luciferase Assay System with Reporter Lysis Buffer (#PAE4030), developed for reporter quantitation in mammalian cells.
- Mild lysis agent and requires a single freeze-thaw cycle to achieve complete cell lysis
- Compatible with two reporter enzyme assays: β-galactosidase (β-gal) and luciferase
The Reporter Lysis Buffer allows β-gal and luciferase assays to be performed on cell lysates of cells cotransfected with vectors carrying these reporter genes.