Order Entry
Puerto Rico
ContactUsLinkComponent
Water ASTM Type III
Water ASTM Type III
Catalog # RCR9151300-4A
Supplier:  Ricca Chemical
   Safer Human and Environmental Health
CAS Number:  
undefined
Water ASTM Type III
Catalog # RCR9151300-4A
Supplier:  Ricca Chemical
Supplier Number:  R9151300-4A
   Safer Human and Environmental Health
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Poly bottle
  • Size:
    4 L
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental Certification:
    EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
  • Cat. No.:
    RCR9151300-4A

Specifications

Specification Test Results

  • Appearance:
    Colorless liquid
  • Chloride (Cl):
    max 10 ppb
  • Conductivity at 25°C:
    max 0.25 µS/cm
  • Organic Carbon (TOC):
    max 200 ppb
  • Silicate (as SiO₂):
    max 500 ppb
  • Sodium (Na):
    max 10 ppb

Specification Test Results

About this item

 :
 :18.02 g/mol
Boiling Pt:100 °C
Melting Pt:0 °C
Density:0.997 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332