Transcription Optimized 5X Buffer, Promega
Catalog # PAP1181
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:Transcription Optimized 5X Buffer
- Size:200 microl
- Storage Conditions:-30°C to -10°C
- Cat. No.:PAP1181
- Supplier no.:P1181
Specifications
About this item
DNA-dependent RNA polymerase for in vitro transcription.
- DNA-Dependent RNA Polymerase Specific for SP6 Promoter Sequences
- Exhibits extremely high affinity and specificity for SP6 promoter sequences
- >90% pure as determined by SDS polyacrylamide gel electrophoresis
- Free of detectable levels of contaminating RNase and DNase activity (
SP6 RNA Polymerase is a DNA-dependent RNA polymerase that exhibits extremely high specificity for its cognate promoter sequences. Only SP6 DNA or DNA cloned downstream from an SP6 promoter can serve as a template for SP6 RNA Polymerase-directed RNA synthesis.