ContactUsLinkComponent
CIAP Buffer Pack, Promega
  PAM1833
 :  
undefined
CIAP Buffer Pack, Promega
  PAM1833
 :  M1833
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    CIAP Buffer Pack
  • Size:
    1.5 mL
  • Cat. No.:
    PAM1833
  • Supplier no.:
    M1833
A complete system for phosphorylating both double- and single-stranded DNA and RNA with T4 PNK and [gamma-32P]ATP.
  • Phosphorylates RNA, ssDNA and dsDNA Ends with a Purchased Radiolabeled Nucleotide
  • System includes enzymes, buffers and control DNA standards for measuring reaction efficiencies (except radionucleotides)
  • Flexible labeling works with [γ-32P]ATP, [γ-33P]ATP or [γ-35S]ATP
The system includes enzymes, buffers and control DNA standards to measure reaction efficiencies. Calf Intestinal Alkaline Phosphatase is included for removal of the 5'-phosphate prior to labeling with T4 Polynucleotide Kinase.
 : Storage Temperature -30°C to -10°C