TE Buffer 20X, (pH 7.5), Promega
Catalog # PAA2651
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:TE Buffer 20X, (pH 7.5)
- Size:25 mL
- Storage Temperature:-30°C to 30°C
- Cat. No.:PAA2651
- Supplier no.:A2651
Specifications
About this item
20X TE Buffer (pH 7.5) is a buffer composed of 0.2mM Tris-HCl and 20mM EDTA (pH 7.5 at 25°C).
- Quality tested and certified free of DNase and RNase activity
- Buffer composition: 0.2mM Tris-HCl, 20mM EDTA (pH 7.5 at 25°C).