Quick-cfDNA™ Serum and Plasma Kit, Zymo Research
Catalog # 77001-374
Supplier: Zymo Research
CAS Number:
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:cfDNA
- Target molecule:DNA
- Description:Quick-cfDNA™ Serum and Plasma Kit
- Size:50 preps
- Description:Quick-cfDNA™ Serum and Plasma Kit
- Cat. No.:77001-374
Specifications
About this item
Quick-cfDNA™ Serum amd Plasma kit provides a simple and reliable method for the rapid preparation of high-quality circulating cell-free DNA from serum, plasma, amniotic fluid, cerebrospinal fluid (CSF), and saliva.
- High-quality DNA, including cell-free, is easily and robustly purified from up to 10 mL of serum, plasma, amniotic fluid or cerebrospinal fluid
- Zymo-Spin™ technology enables elution of DNA in as little as 35 μl
- Ensures it is ready for all sensitive downstream applications such as qPCR and Next-Generation sequencing
A combination of chemical and enzymatic methods are used to efficiently recover total DNA (including cell-free apoptotic, necrotic, mitochondrial and viral DNA) linearly from up to 10 mL of sample.
Delivery: Kit includes Proteinase K, Proteinase K storage buffer, S&P 5X digestion buffer, S&P DNA binding buffer, S&P DNA Prep buffer, S&P DNA wash buffer (concentrate), DNA elution buffer, Zymo-Spin™ III-S column assembly, collection tubes and instruction manual.