



Bromine number titration solvent ASTM 1159 & ASTM D 2710
Catalog # 103193-884
Supplier: Aqua Solutions
CAS Number:
Specifications
- Pack type:Glass, amber
- Size:4 L
- Cat. No.:103193-884
- Supplier no.:SPX191
Specifications
About this item
:
UN:3286
ADR:8,II
:GHS02
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient