Colorimetric Gas Detector Tubes, Sensidyne
Catalog # 13502-630
Supplier: Sensidyne
Small Business Enterprise
Specifications
- Temperature range:0 – 45 °C
- Description:Detector tubes, 1 min (50 ml) or 1.5 min (100 ml)
- Cat. No.:13502-630
- Measurement range:3–24% (50 ml), 1–3% (100 ml)
- No. of strokes:½ (50 ml) or 1 (100 ml)
- Detectable gas:Oxygen
- Supplier no.:159SC
Specifications
About this item
Detector tubes for detection of oxygen-non-heating type.
- Detector tubes are easy to use
- Best when sampling for oxygen-non-heating type or numerous other gases and vapors
- Color changes black to white
- Direct reading from the scale calibrated by 1/2 pump stroke
- Shelf life 2 years
This allows users to perform field screening and tests for the presence of hazardous and toxic gases, and vapours at a very low cost-per-sample. When the concentration is below 3%, 1 pump stroke can be used to determine the lower concentration.