ContactUsLinkComponent
Disposable Magnetic Pad, CARDINAL HEALTH
Disposable Magnetic Pad, CARDINAL HEALTH
Catalog # PH200-16A
Supplier:  Cardinal Health
Disposable Magnetic Pad, CARDINAL HEALTH
Catalog # PH200-16A
Supplier:  Cardinal Health
Supplier Number:  200-16B
Specifications
  • Sterility:
    Sterile
  • Description:
    Disposable Magnetic Instrument Pad
  • Dimensions:
    10×16"
  • Cat. No.:
    PH200-16A
  • Supplier no.:
    200-16B
Specifications
About this item
Disposable magnetic pad covered with antistatic polyethylene.
It has lightweight drapes have multiple raised magnetic elements on a nonslip foam back. Allow positive instrument control adjacent to the surgical site.