Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD6 Mouse Monoclonal Antibody (CF405S) [clone: C6/372 3F7B5]
Anti-CD6 Mouse Monoclonal Antibody (CF405S) [clone: C6/372 3F7B5]
Supplier:  Biotium
Certificates
Anti-CD6 Mouse Monoclonal Antibody (CF405S) [clone: C6/372 3F7B5]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75918-548
  • 75916-536
  • Antigen Symbol:
  • CD6
  • CD6
  • Antigen Name:
  • CD6
  • CD6
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • C6/372 3F7B5
  • C6/372 3F7B5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 744366
  • Unigene ID: 744366
  • Isotype:
  • IgGs
  • IgGs
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together