Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-S100A1 Mouse Monoclonal Antibody (CF594) [clone: 4C4.9 S100A1/1012]
 :  Biotium
 
Anti-S100A1 Mouse Monoclonal Antibody (CF594) [clone: 4C4.9 S100A1/1012]
 :  Biotium

 

  • Catalog No:
  • 75975-594
  • 75975-592
  • Antigen Symbol:
  • S100A1
  • S100A1
  • Antigen Name:
  • S100
  • S100
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • 4C4.9 S100A1/1012
  • 4C4.9 S100A1/1012
  • Reactivity:
  • Human, Bovine, Rat, Mouse
  • Human, Bovine, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 515715
  • Unigene ID: 515715
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide