Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PTH Mouse Monoclonal Antibody (CF640R) [clone: PTH/1175]
Anti-PTH Mouse Monoclonal Antibody (CF640R) [clone: PTH/1175]
Supplier:  Biotium
Certificates
Anti-PTH Mouse Monoclonal Antibody (CF640R) [clone: PTH/1175]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75973-486
  • 75973-484
  • Antigen Symbol:
  • PTH
  • PTH
  • Antigen Name:
  • PTH / Parathyroid Hormone
  • PTH / Parathyroid Hormone
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • PTH/1175
  • PTH/1175
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 37045
  • Unigene ID: 37045
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications