Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-WT1 Mouse Monoclonal Antibody (CF568) [clone: WT1/857 6F-H2]
Anti-WT1 Mouse Monoclonal Antibody (CF568) [clone: WT1/857 6F-H2]
Supplier:  Biotium
Certificates
Anti-WT1 Mouse Monoclonal Antibody (CF568) [clone: WT1/857 6F-H2]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75980-042
  • 75980-040
  • Antigen Symbol:
  • WT1
  • WT1
  • Antigen Name:
  • WT1
  • WT1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • WT1/857 6F-H2
  • WT1/857 6F-H2
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 591980
  • Unigene ID: 591980
  • Isotype:
  • IgG
  • IgG
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together